Thermo Scientific Chemicals

3-Bromobenzhydrazide, 98+%, Thermo Scientific Chemicals

Catalog number: A19628.14
25 g, Each
Thermo Scientific Chemicals

3-Bromobenzhydrazide, 98+%, Thermo Scientific Chemicals

Catalog number: A19628.14
25 g, Each
Quantity
Catalog number: A19628.14
also known as A19628-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39115-96-3
IUPAC Name
3-bromobenzohydrazide
Molecular Formula
C7H7BrN2O
InChI Key
BNAQRAZIPAHWAR-UHFFFAOYSA-N
SMILES
NNC(=O)C1=CC=CC(Br)=C1
Appearance (Color)
White to pale cream to pale brown
Form
Powder
Assay (GC)
≥98.0%
Melting Point (clear melt)
153.0-159.0?C

Description

3-Bromobenzhydrazide is used to produce 5-(3-bromo-phenyl)-3H-[1,3,4]oxadiazole-2-thione. This reaction will need reagent potassium hydroxide and solvent ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzhydrazide is used to produce 5-(3-bromo-phenyl)-3H-[1,3,4]oxadiazole-2-thione. This reaction will need reagent potassium hydroxide and solvent ethanol.

Solubility
Slightly soluble in water (3.7 g/L) (25°C)

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text