Thermo Scientific Chemicals

N,N-Diethylethylenediamine, 99%, Thermo Scientific Chemicals

Catalog number: A19636.22
100 g, Each
Thermo Scientific Chemicals

N,N-Diethylethylenediamine, 99%, Thermo Scientific Chemicals

Catalog number: A19636.22
100 g, Each
Quantity
Catalog number: A19636.22
also known as A19636-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-36-7
IUPAC Name
(2-aminoethyl)diethylamine
Molecular Formula
C6H16N2
InChI Key
UDGSVBYJWHOHNN-UHFFFAOYSA-N
SMILES
CCN(CC)CCN
Identification (FTIR)
Conforms
Refractive Index
1.4340-1.4370 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Form
Liquid

Description

N,N-Diethylethylenediamine was used in the oxidative amidation of hydrogen phosphonate diesters in the synthesis of oligonucleotides. It is also used to develop melanin targeted PET and single-photon emission computed tomography (SPECT) imaging agents for melanoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylethylenediamine was used in the oxidative amidation of hydrogen phosphonate diesters in the synthesis of oligonucleotides. It is also used to develop melanin targeted PET and single-photon emission computed tomography (SPECT) imaging agents for melanoma.

Solubility
Miscible in water.

Notes
Air sensitive. Store away from oxidizing agents. Protect against electrostatic charges. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text