Thermo Scientific Chemicals

Isovaleraldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A19640.AE
100 mL, Each
Thermo Scientific Chemicals

Isovaleraldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A19640.AE
100 mL, Each
Quantity
Catalog number: A19640.AE
also known as A19640-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
590-86-3
IUPAC Name
3-methylbutanal
Molecular Formula
C5H10O
InChI Key
YGHRJJRRZDOVPD-UHFFFAOYSA-N
SMILES
CC(C)CC=O
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Free acid (titration)
≤1.5%

Description

Isovaleraldehyde acts as a reagent in the preparation of active pharmaceutical ingredient (API) products. It serves as an internal standard for the determination of wine aroma carbonyl compounds with 5-8 carbon atoms. Further, it is utilized as a standard to evaluate the quality of olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis. In addition to this, it is employed to enhance the taste and odor of the food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isovaleraldehyde acts as a reagent in the preparation of active pharmaceutical ingredient (API) products. It serves as an internal standard for the determination of wine aroma carbonyl compounds with 5-8 carbon atoms. Further, it is utilized as a standard to evaluate the quality of olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis. In addition to this, it is employed to enhance the taste and odor of the food.

Solubility
Miscible with alcohol and diethyl ether. Slightly miscible with water.

Notes
Light and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, bases, alkaline materials and reducing agents. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text