Phenanthrene is generally used for dye production, synthetic resin, preservative. It is also used in agrochemical and pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenanthrene is generally used for dye production, synthetic resin, preservative. It is also used in agrochemical and pharmaceutical intermediates.
Solubility
Soluble in alcohol, benzene, toluene, and glacial acetic acid
Notes
Light sensitive. Handle and store under inert gas. Incompatible with strong oxidizing agents.
RUO – Research Use Only
T. Azumi and S. P. McGlynn. Polarization of the Luminescence of Phenanthrene.J. Chem. Phys.1962, 37 2413.
Yanzheng Gao; Lizhong Zhu. Plant uptake, accumulation and translocation of phenanthrene and pyrene in soils.Chemosphere.2004, 55 1169-1178.