3H-Tetrafluoropropionic acid, 97%, Thermo Scientific Chemicals
3H-Tetrafluoropropionic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3H-Tetrafluoropropionic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
Catalog number A19648.09
also known as A19648-09
Price (USD)/ Each
210.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
210.00
Add to cart
3H-Tetrafluoropropionic acid, 97%, Thermo Scientific Chemicals
Catalog numberA19648.09
Price (USD)/ Each
210.00
-
Add to cart
Chemical Identifiers
CAS756-09-2
IUPAC Name2,2,3,3-tetrafluoropropanoate
Molecular FormulaC3HF4O2
InChI KeyPXRROZVNOOEPPZ-UHFFFAOYSA-M
SMILES[O-]C(=O)C(F)(F)C(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (GC)≥96.0%
Refractive Index1.3205-1.3255 @ 20°C
FormLiquid or fuming liquid
3H-Tetrafluoropropionic acid is used to produce 2,2,3,3-tetrafluoro-propionyl chloride. This reaction will need reagent benzotrichloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3H-Tetrafluoropropionic acid is used to produce 2,2,3,3-tetrafluoro-propionyl chloride. This reaction will need reagent benzotrichloride.

Solubility
Sparingly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only