Thermo Scientific Chemicals

2-Methylbutyraldehyde, 95%, Thermo Scientific Chemicals

Catalog number: A19650.22
100 g, Each
Thermo Scientific Chemicals

2-Methylbutyraldehyde, 95%, Thermo Scientific Chemicals

Catalog number: A19650.22
100 g, Each
Quantity
Catalog number: A19650.22
also known as A19650-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-17-3
IUPAC Name
2-methylbutanal
Molecular Formula
C5H10O
InChI Key
BYGQBDHUGHBGMD-UHFFFAOYNA-N
SMILES
CCC(C)C=O
Assay (GC)
≥94.0%
Refractive Index
1.3890-1.3950 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Form
Liquid

Description

2-Methylbutyraldehyde is used for lift in woody and cocoa topnote complexes. And also used in cocoa, coffee, wine and nut flavors for lift.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbutyraldehyde is used for lift in woody and cocoa topnote complexes. And also used in cocoa, coffee, wine and nut flavors for lift.

Solubility
Soluble in water, ether, and alcohol.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text