Thermo Scientific Chemicals

N,N-Dimethylglycine, 98+%, Thermo Scientific Chemicals

Catalog number: A19670.22
100 g, Each
Thermo Scientific Chemicals

N,N-Dimethylglycine, 98+%, Thermo Scientific Chemicals

Catalog number: A19670.22
100 g, Each
Quantity
Catalog number: A19670.22
also known as A19670-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1118-68-9
IUPAC Name
2-(dimethylamino)acetic acid
Molecular Formula
C4H9NO2
InChI Key
FFDGPVCHZBVARC-UHFFFAOYSA-N
SMILES
CN(C)CC(O)=O
Melting Point
175-183°C
Appearance (Color)
White to cream or pale yellow
Form
Crystalline powder or powder
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Water Content (Karl Fischer Titration)
≤1.75%

Description

N,N-Dimethylglycine is used as an athletic performance enhancer and immunostimulant. It is also used in the treatment of autism, epilepsy and mitochondrial disease. It is also employed as a biomarker of protein degradation in chronic obstructive lung disease (COPD) patients. Further, it is used as a substrate to identify, differentiate and characterize amino acid methyltransferase. It plays an important role for the development of glycine-based ionic liquids and emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text