Thermo Scientific Chemicals

4-Fluoro-3-nitrobenzotrifluoride, 97%, Thermo Scientific Chemicals

Catalog number: A19676.22
100 g, Each
Thermo Scientific Chemicals

4-Fluoro-3-nitrobenzotrifluoride, 97%, Thermo Scientific Chemicals

Catalog number: A19676.22
100 g, Each
Quantity
Catalog number: A19676.22
also known as A19676-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
367-86-2
IUPAC Name
1-fluoro-2-nitro-4-(trifluoromethyl)benzene
Molecular Formula
C7H3F4NO2
InChI Key
HLDFCCHSOZWKAA-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC(=CC=C1F)C(F)(F)F
Assay (GC)
≥96.0%
Refractive Index
1.4595-1.4645 @ 20?C
Appearance (Color)
Clear yellow to orange
Form
Liquid

Description

4-Fluoro-3-nitrobenzotrifluoride is used in the pre-column derivatization technique for HPLC with UV/VIS spectrophotometric detection of polyamines. It may be used as derivatization reagent for the HPLC determination of polyamines. It was also used in the synthesis of 2-nitro-4-trifluoromethylphenylhydrazine, derivatization reagent for 1,2-diol compounds and oxo-steroids.
  • [3-(2-nitro-4-trifluoromethylphenyl)aminophenyl]dihydroxyborane, derivatization reagent for 1,2-diol compounds and oxo-steroids3
  • 1-(3-chlorophenyl)-5-trifluoromethyl-3-hydrobenzimidazol-2-one

    This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

    Applications
    4-Fluoro-3-nitrobenzotrifluoride is used in the pre-column derivatization technique for HPLC with UV/VIS spectrophotometric detection of polyamines. It may be used as derivatization reagent for the HPLC determination of polyamines. It was also used in the synthesis of 2-nitro-4-trifluoromethylphenylhydrazine, derivatization reagent for 1,2-diol compounds and oxo-steroids. • [3-(2-nitro-4-trifluoromethylphenyl)aminophenyl]dihydroxyborane, derivatization reagent for 1,2-diol compounds and oxo-steroids3 • 1-(3-chlorophenyl)-5-trifluoromethyl-3-hydrobenzimidazol-2-one

    Solubility
    Insoluble in water.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
    RUO – Research Use Only

    Figures

    Documents & Downloads

    Certificates

      Frequently asked questions (FAQs)

      Citations & References

      Search citations by name, author, journal title or abstract text