1,3-Dimethyladamantane, 98%, Thermo Scientific Chemicals
1,3-Dimethyladamantane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Dimethyladamantane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A19684.14
also known as A19684-14
Price (USD)/ Each
241.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
241.00
Add to cart
1,3-Dimethyladamantane, 98%, Thermo Scientific Chemicals
Catalog numberA19684.14
Price (USD)/ Each
241.00
-
Add to cart
Chemical Identifiers
CAS702-79-4
IUPAC Name1,3-dimethyladamantane
Molecular FormulaC12H20
InChI KeyCWNOIUTVJRWADX-UHFFFAOYSA-N
SMILESCC12CC3CC(C1)CC(C)(C3)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4760-1.4800 @ 20?C
Assay (GC)≥97.5%
FormLiquid
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only