Thermo Scientific Chemicals

4-Hydroxy-6-methyl-3-nitro-2-pyridone, 97%, Thermo Scientific Chemicals

Catalog number: A19690.14
25 g, Each
Thermo Scientific Chemicals

4-Hydroxy-6-methyl-3-nitro-2-pyridone, 97%, Thermo Scientific Chemicals

Catalog number: A19690.14
25 g, Each
Quantity
Catalog number: A19690.14
also known as A19690-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4966-90-9
IUPAC Name
2-hydroxy-6-methyl-3-nitro-1,4-dihydropyridin-4-one
Molecular Formula
C6H6N2O4
InChI Key
QIKWTNPFTOEELW-UHFFFAOYSA-N
SMILES
CC1=CC(=O)C(=C(O)N1)[N+]([O-])=O
Appearance (Color)
White to yellow
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to yellow

Description

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Solubility in N,N-DMF gives very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text