Thermo Scientific Chemicals

Ethyl thiophene-3-acetate, 98%, Thermo Scientific Chemicals

Catalog number: A19694.14
25 g, Each
Thermo Scientific Chemicals

Ethyl thiophene-3-acetate, 98%, Thermo Scientific Chemicals

Catalog number: A19694.14
25 g, Each
Quantity
Catalog number: A19694.14
also known as A19694-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
37784-63-7
IUPAC Name
ethyl 2-(thiophen-3-yl)acetate
Molecular Formula
C8H10O2S
InChI Key
FZBNQIWPYCUPAP-UHFFFAOYSA-N
SMILES
CCOC(=O)CC1=CSC=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Refractive Index
1.5080-1.5120 @ 20?C
Assay (GC)
≥97.5%

Description

Ethyl thiophene-3-acetate is used in the syntheses of poly(thiophene-3-acetic acid) and solution-processable polymers, which consist of polythiophene main chain and alkyl and oligoaniline side groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl thiophene-3-acetate is used in the syntheses of poly(thiophene-3-acetic acid) and solution-processable polymers, which consist of polythiophene main chain and alkyl and oligoaniline side groups.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text