Thermo Scientific Chemicals

4-Amino-3-chlorophenol hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A19720.22
100 g, Each
Thermo Scientific Chemicals

4-Amino-3-chlorophenol hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A19720.22
100 g, Each
Quantity
Catalog number: A19720.22
also known as A19720-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52671-64-4
IUPAC Name
4-amino-3-chlorophenol
Molecular Formula
C6H6ClNO
InChI Key
PNLPXABQLXSICH-UHFFFAOYSA-N
SMILES
NC1=CC=C(O)C=C1Cl
Melting Point
ca 250-255?C dec.
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Appearance (Color)
Pale cream to pale grey or pale brown
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text