Ethyl 4,4-dimethyl-3-oxovalerate, 97%, Thermo Scientific Chemicals
Ethyl 4,4-dimethyl-3-oxovalerate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 4,4-dimethyl-3-oxovalerate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number A19752.22
also known as A19752-22
Price (USD)/ Each
234.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
234.00
Add to cart
Ethyl 4,4-dimethyl-3-oxovalerate, 97%, Thermo Scientific Chemicals
Catalog numberA19752.22
Price (USD)/ Each
234.00
-
Add to cart
Chemical Identifiers
CAS17094-34-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4295-1.4345 @ 20?C
FormLiquid
Assay (GC)≥96.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only