Thermo Scientific Chemicals

2,2,4-Trimethylpentane, 99%, Thermo Scientific Chemicals

Catalog number: A19756.0F
2500 mL, Each
Thermo Scientific Chemicals

2,2,4-Trimethylpentane, 99%, Thermo Scientific Chemicals

Catalog number: A19756.0F
2500 mL, Each
Quantity
Catalog number: A19756.0F
also known as A19756-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
540-84-1
IUPAC Name
2,2,4-trimethylpentane
Molecular Formula
C8H18
InChI Key
NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES
CC(C)CC(C)(C)C
Assay (GC)
≥98.5%
Refractive Index
1.3900-1.3930 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms

Description

2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and in organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as thinner. It is involved as processing aids in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and in organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as thinner. It is involved as processing aids in the petroleum industry.

Solubility
Miscible with benzene, toluene, xylene, chloroform, ether, carbon disulfide, carbon tetrachloride, DMF, acetone, heptane, and oils. Immiscible with water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from sources of ignition. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text