Thermo Scientific Chemicals

2,4,6-Trifluorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A19758.03
1 g, Each
Thermo Scientific Chemicals

2,4,6-Trifluorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A19758.03
1 g, Each
Quantity
Catalog number: A19758.03
also known as A19758-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28314-80-9
IUPAC Name
2,4,6-trifluorobenzoate
Molecular Formula
C7H2F3O2
InChI Key
SJZATRRXUILGHH-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1=C(F)C=C(F)C=C1F
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
141.0-148.0?C
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text