Thermo Scientific Chemicals

Farnesyl acetate, mixture of isomers, 96%, Thermo Scientific Chemicals

Catalog number: A19778.22
100 g, Each
Thermo Scientific Chemicals

Farnesyl acetate, mixture of isomers, 96%, Thermo Scientific Chemicals

Catalog number: A19778.22
100 g, Each
Quantity
Catalog number: A19778.22
also known as A19778-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29548-30-9
IUPAC Name
(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trien-1-yl acetate
Molecular Formula
C17H28O2
InChI Key
ZGIGZINMAOQWLX-NCZFFCEISA-N
SMILES
CC(C)=CCC\C(C)=C\CC\C(C)=C\COC(C)=O
Form
Liquid
Refractive Index
1.4750-1.4810 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥95.0% (sum of isomers)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text