4-Bromo-2-chloro-1-fluorobenzene, 98+%, Thermo Scientific Chemicals
4-Bromo-2-chloro-1-fluorobenzene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-2-chloro-1-fluorobenzene, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number A19782.22
also known as A19782-22
Price (USD)/ Each
692.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
692.00
Add to cart
4-Bromo-2-chloro-1-fluorobenzene, 98+%, Thermo Scientific Chemicals
Catalog numberA19782.22
Price (USD)/ Each
692.00
-
Add to cart
Chemical Identifiers
CAS60811-21-4
IUPAC Name4-bromo-2-chloro-1-fluorobenzene
Molecular FormulaC6H3BrClF
InChI KeyCJTIWGBQCVYTQE-UHFFFAOYSA-N
SMILESFC1=C(Cl)C=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
Refractive Index1.5520-1.5560 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only