4,5-Dibromothiophene-2-carboxaldehyde, 98%, Thermo Scientific Chemicals
4,5-Dibromothiophene-2-carboxaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,5-Dibromothiophene-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A19788.14
also known as A19788-14
Price (USD)/ Each
486.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
486.00
Add to cart
4,5-Dibromothiophene-2-carboxaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberA19788.14
Price (USD)/ Each
486.00
-
Add to cart
Chemical Identifiers
CAS38071-22-6
IUPAC Name4,5-dibromothiophene-2-carbaldehyde
Molecular FormulaC5H2Br2OS
InChI KeyCHRAVDVWAPGZOY-UHFFFAOYSA-N
SMILESBrC1=C(Br)C=C(S1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
Assay (GC)≥97.5%
FormPowder
Melting Point (clear melt)77.5-83.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only