Thermo Scientific Chemicals

4-Methylmorpholine N-oxide, 50% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: A19802.0B
1000 g, Each
Thermo Scientific Chemicals

4-Methylmorpholine N-oxide, 50% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: A19802.0B
1000 g, Each
Quantity
Catalog number: A19802.0B
also known as A19802-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7529-22-8
IUPAC Name
4-methylmorpholin-4-ium-4-olate
Molecular Formula
C5H11NO2
InChI Key
LFTLOKWAGJYHHR-UHFFFAOYSA-N
SMILES
C[N+]1([O-])CCOCC1
Appearance (Color)
Clear colorless to pale yellow
Assay (Non-aqueous acid-base Titration)
≥49.0 to ≤52.0%
Refractive Index
1.4190-1.4250 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms

Description

4-Methylmorpholine N-oxide acts as a non-metallic catalyst for the cyanosilylation of ketones. It is also employed as a co-oxidant for Sharpless asymmetric dihydroxylation in ionic liquids. It serves as a solvent in the Lyocell process to produce tencel fiber. Further, it is used in the preparation of aldehydes from primary alcohols in the presence of tetrapropylammonium perruthenate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylmorpholine N-oxide acts as a non-metallic catalyst for the cyanosilylation of ketones. It is also employed as a co-oxidant for Sharpless asymmetric dihydroxylation in ionic liquids. It serves as a solvent in the Lyocell process to produce tencel fiber. Further, it is used in the preparation of aldehydes from primary alcohols in the presence of tetrapropylammonium perruthenate.

Solubility
Soluble in water, methanol, ethanol, acetone, ethers and dimethylsulfoxide.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides and chloroformates.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text