4-Ethoxybenzonitrile, 98%, Thermo Scientific Chemicals
4-Ethoxybenzonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Ethoxybenzonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A19806.06
also known as A19806-06
Price (USD)/ Each
48.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
48.20
Add to cart
4-Ethoxybenzonitrile, 98%, Thermo Scientific Chemicals
Catalog numberA19806.06
Price (USD)/ Each
48.20
-
Add to cart
Chemical Identifiers
CAS25117-74-2
IUPAC Name4-ethoxybenzonitrile
Molecular FormulaC9H9NO
InChI KeyPJRLUGQMEZZDIY-UHFFFAOYSA-N
SMILESCCOC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only