Nerol, 97%, Thermo Scientific Chemicals
Nerol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nerol, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 mL
Catalog number A19808.AE
also known as A19808-AE
Price (USD)/ Each
43.65
Online exclusive
48.00 
Save 4.35 (9%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
43.65
Online exclusive
48.00 
Save 4.35 (9%)
Add to cart
Nerol, 97%, Thermo Scientific Chemicals
Catalog numberA19808.AE
Price (USD)/ Each
43.65
Online exclusive
48.00 
Save 4.35 (9%)
-
Add to cart
Chemical Identifiers
CAS106-25-2
IUPAC Name(2Z)-3,7-dimethylocta-2,6-dien-1-ol
Molecular FormulaC10H18O
InChI KeyGLZPCOQZEFWAFX-YFHOEESVSA-N
SMILESCC(C)=CCC\C(C)=C/CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Refractive Index1.4720-1.4790 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only