Thermo Scientific Chemicals

Cyclohexanecarbonyl chloride, 97+%, Thermo Scientific Chemicals

Catalog number: A19824.22
100 g, Each
Thermo Scientific Chemicals

Cyclohexanecarbonyl chloride, 97+%, Thermo Scientific Chemicals

Catalog number: A19824.22
100 g, Each
Quantity
Catalog number: A19824.22
also known as A19824-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2719-27-9
IUPAC Name
cyclohexanecarbonyl chloride
Molecular Formula
C7H11ClO
InChI Key
RVOJTCZRIKWHDX-UHFFFAOYSA-N
SMILES
ClC(=O)C1CCCCC1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.0%
Refractive Index
1.4670-1.4720 @ 20°C (non-U.S. specification)

Description

Cyclohexanecarbonyl chloride is used in the preparation of five thiourea derivative ligands having anti-bacterial and anti-yeast activity, (N-(diethylcarbamothioyl)cyclohexanecarboxamide, N-(di-n-propylcarbamothioyl)cyclohexanecarboxamide, di-n-butylcarbamothioyl)cyclohexanecarboxamide, N-(diphenylcarbamothioyl)cyclohexanecarboxamide, N-(morpholine-4-carbonothioyl)cyclohexanecarboxamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanecarbonyl chloride is used in the preparation of five thiourea derivative ligands having anti-bacterial and anti-yeast activity, (N-(diethylcarbamothioyl)cyclohexanecarboxamide, N-(di-n-propylcarbamothioyl)cyclohexanecarboxamide, di-n-butylcarbamothioyl)cyclohexanecarboxamide, N-(diphenylcarbamothioyl)cyclohexanecarboxamide, N-(morpholine-4-carbonothioyl)cyclohexanecarboxamide.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text