3-Nonanone, 98%, Thermo Scientific Chemicals
3-Nonanone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Nonanone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number A19878.22
also known as A19878-22
Price (USD)/ Each
101.65
Online exclusive
113.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
101.65
Online exclusive
113.00 
Save 11.35 (10%)
Add to cart
3-Nonanone, 98%, Thermo Scientific Chemicals
Catalog numberA19878.22
Price (USD)/ Each
101.65
Online exclusive
113.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS925-78-0
IUPAC Namenonan-3-one
Molecular FormulaC9H18O
InChI KeyIYTXKIXETAELAV-UHFFFAOYSA-N
SMILESCCCCCCC(=O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4185-1.4225 @ 20?C
FormLiquid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only