Thermo Scientific Chemicals

2,5-Dimethoxybenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: A19928.14
25 g, Each
Thermo Scientific Chemicals

2,5-Dimethoxybenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: A19928.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A19928.14
also known as A19928-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-02-7
IUPAC Name
2,5-dimethoxybenzaldehyde
Molecular Formula
C9H10O3
InChI Key
AFUKNJHPZAVHGQ-UHFFFAOYSA-N
SMILES
COC1=CC=C(OC)C(C=O)=C1
Form
Crystals or powder or crystalline powder or chunks or flakes
Assay (GC)
≥98.0%
Melting Point (clear melt)
45.0-51.0?C
Appearance (Color)
Pale cream to cream to yellow to pale brown
Free acid (titration)
≤1%

Description

2,5-Dimethoxybenzaldehyde is used in the preparation of 2,5-dimethoxyphenethylamine, which is utilized to prepare psychoactive drugs such as 2,5-dimethoxy-4-bromophenethylamine, 2,5-dimethoxy-4-iodophenethylamine and 4-Chloro-2,5-dimethoxy-phenethylamine. It acts as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethoxybenzaldehyde is used in the preparation of 2,5-dimethoxyphenethylamine, which is utilized to prepare psychoactive drugs such as 2,5-dimethoxy-4-bromophenethylamine, 2,5-dimethoxy-4-iodophenethylamine and 4-Chloro-2,5-dimethoxy-phenethylamine. It acts as an intermediate in organic synthesis.

Solubility
Soluble in chloroform and methanol. Slightly soluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text