Thermo Scientific Chemicals

Sodium tert-butoxide, 97%, Thermo Scientific Chemicals

Catalog number: A19942.22
100 g, Each
Thermo Scientific Chemicals

Sodium tert-butoxide, 97%, Thermo Scientific Chemicals

Catalog number: A19942.22
100 g, Each
Quantity
Catalog number: A19942.22
also known as A19942-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
865-48-5
IUPAC Name
sodium 2-methylpropan-2-olate
Molecular Formula
C4H9NaO
InChI Key
MFRIHAYPQRLWNB-UHFFFAOYSA-N
SMILES
[Na+].CC(C)(C)[O-]
Impurity content
Sodium hydroxide and sodium carbonate (as hydroxide): <2.0%
Appearance (Color)
White to cream to pale brown
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder

Description

Sodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.

Notes
Air sensitive and hygroscopic. Incompatible with acids, reducing agents, oxygen, water, alcohols, chlorinated solvents and halogens.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text