Thermo Scientific Chemicals

Zinc selenide, Thermo Scientific Chemicals

Catalog number: A19947.09
10 g, Each
Thermo Scientific Chemicals

Zinc selenide, Thermo Scientific Chemicals

Catalog number: A19947.09
10 g, Each
Quantity
Catalog number: A19947.09
also known as A19947-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1315-09-9
IUPAC Name
zinc(2+) selandiide
Molecular Formula
SeZn
InChI Key
GTLQJUQHDTWYJC-UHFFFAOYSA-N
SMILES
[Zn++].[Se--]
Form
powder
Appearance (Color)
orange or red to brown
Complexometric Titration
> 88% by zinc

Description

Zinc selenide is used to form II-VI light-emitting diodes and diode lasers. It is doped with chromium (ZnSe:Cr) has been used as an infrared laser gain medium. It is used as an infrared optical material. It is used in x-ray and gamma ray detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc selenide is used to form II-VI light-emitting diodes and diode lasers. It is doped with chromium (ZnSe:Cr) has been used as an infrared laser gain medium. It is used as an infrared optical material. It is used in x-ray and gamma ray detectors.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text