Thermo Scientific Chemicals

2-Methyl-4-octanone, 99%, Thermo Scientific Chemicals

Catalog number: A19952.14
25 g, Each
Thermo Scientific Chemicals

2-Methyl-4-octanone, 99%, Thermo Scientific Chemicals

Catalog number: A19952.14
25 g, Each
Quantity
Catalog number: A19952.14
also known as A19952-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7492-38-8
IUPAC Name
2-methyloctan-4-one
Molecular Formula
C9H18O
InChI Key
ICSKJDZASFIJQK-UHFFFAOYSA-N
SMILES
CCCCC(=O)CC(C)C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.4145-1.4175 @ 20?C

Description

2-Methyl-4-octanone is a building block ketone for proteomics research. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-4-octanone is a building block ketone for proteomics research. It is also used as pharmaceutical intermediates.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text