Thermo Scientific™

Triisopropanolamine, 95%, Thermo Scientific Chemicals

Catalog number: A19966.36
500 g, Each
Thermo Scientific™

Triisopropanolamine, 95%, Thermo Scientific Chemicals

Catalog number: A19966.36
500 g, Each
Quantity
Catalog number: A19966.36
also known as A19966-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122-20-3
IUPAC Name
1-[bis(2-hydroxypropyl)amino]propan-2-ol
Molecular Formula
C9H21NO3
InChI Key
SLINHMUFWFWBMU-UHFFFAOYNA-N
SMILES
CC(O)CN(CC(C)O)CC(C)O
Appearance (Color)
White to yellow
Form
Fused solid or clear liquid as melt
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0%
Identification (FTIR)
Conforms

Description

Major applications include water-based coating applications and agricultural products. Additional applications are antistat agents for polymers, corrosion inhibitor, electrodeposition/electrocoating, lubricants, paper, pigment dispersion, plastics, polyurethane additive, reaction intermediates, rubber curing, surfactants, mineral dispersion, and urethanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Major applications include water-based coating applications and agricultural products. Additional applications are antistat agents for polymers, corrosion inhibitor, electrodeposition/electrocoating, lubricants, paper, pigment dispersion, plastics, polyurethane additive, reaction intermediates, rubber curing, surfactants, mineral dispersion, and urethanes.

Solubility
Insoluble in water. Soluble ethanol, diethyl ether, chloroform (slightly ), and methanol (>500 g/100g).

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text