Cyclopentyl phenyl ketone, 96%, Thermo Scientific Chemicals
Cyclopentyl phenyl ketone, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopentyl phenyl ketone, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A19976.03
also known as A19976-03
Price (USD)/ Each
34.60
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
34.60
Add to cart
Cyclopentyl phenyl ketone, 96%, Thermo Scientific Chemicals
Catalog numberA19976.03
Price (USD)/ Each
34.60
-
Add to cart
Chemical Identifiers
CAS5422-88-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to yellow
FormLiquid
Assay (GC)≥95.0%
Refractive Index1.5415-1.5475 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only