Ethyl 2-nitrobenzoate, 97%, Thermo Scientific Chemicals
Ethyl 2-nitrobenzoate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-nitrobenzoate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A19994.14
also known as A19994-14
Price (USD)/ Each
67.10
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
67.10
Add to cart
Ethyl 2-nitrobenzoate, 97%, Thermo Scientific Chemicals
Catalog numberA19994.14
Price (USD)/ Each
67.10
-
Add to cart
Chemical Identifiers
CAS610-34-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow or yellow-orange
CommentMay contain some dark coloured particles
FormCrystals, Crystalline powder or fused solid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only