Sodium 2-chloroethanesulfonate hydrate, 98+% (dry wt.), water <10%, Thermo Scientific Chemicals
Sodium 2-chloroethanesulfonate hydrate, 98+% (dry wt.), water &lt;10%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium 2-chloroethanesulfonate hydrate, 98+% (dry wt.), water <10%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A19995.22
also known as A19995-22
Price (USD)/ Each
425.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
425.00
Add to cart
Sodium 2-chloroethanesulfonate hydrate, 98+% (dry wt.), water <10%, Thermo Scientific Chemicals
Catalog numberA19995.22
Price (USD)/ Each
425.00
-
Add to cart
Specifications
Chemical Name or MaterialSodium 2-chloroethanesulfonate hydrate
CAS15484-44-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only