Thermo Scientific Chemicals

3-Methyl-2-pentanone, 98+%, Thermo Scientific Chemicals

Catalog number: A19999.09
10 g, Each
Thermo Scientific Chemicals

3-Methyl-2-pentanone, 98+%, Thermo Scientific Chemicals

Catalog number: A19999.09
10 g, Each
Quantity
Catalog number: A19999.09
also known as A19999-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
565-61-7
IUPAC Name
3-methylpentan-2-one
Molecular Formula
C6H12O
InChI Key
UIHCLUNTQKBZGK-UHFFFAOYNA-N
SMILES
CCC(C)C(C)=O
Appearance (Color)
Clear colorless
Assay (GC)
≥98.0%
Refractive Index
1.3980-1.4020 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

Description

3-Methyl-2-pentanone is used as a solvent and as an intermediate for syntheses of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-pentanone is used as a solvent and as an intermediate for syntheses of pharmaceuticals.

Solubility
Soluble in alcohol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text