3-Methyl-2-pentanone, 98+%, Thermo Scientific Chemicals
3-Methyl-2-pentanone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methyl-2-pentanone, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
25 g
Catalog number A19999.18
also known as A19999-18
Price (USD)/ Each
352.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
352.00
Add to cart
3-Methyl-2-pentanone, 98+%, Thermo Scientific Chemicals
Catalog numberA19999.18
Price (USD)/ Each
352.00
-
Add to cart
Chemical Identifiers
CAS565-61-7
IUPAC Name3-methylpentan-2-one
Molecular FormulaC6H12O
InChI KeyUIHCLUNTQKBZGK-UHFFFAOYNA-N
SMILESCCC(C)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
Refractive Index1.3980-1.4020 @ 20?C
Identification (FTIR)Conforms
FormLiquid
3-Methyl-2-pentanone is used as a solvent and as an intermediate for syntheses of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-pentanone is used as a solvent and as an intermediate for syntheses of pharmaceuticals.

Solubility
Soluble in alcohol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only