Owl™ A2 Large Gel Systems
Owl™ A2 Large Gel Systems
Thermo Scientific™

Owl™ A2 Large Gel Systems

Handle high throughput with consistent, reliable results with Thermo Scientific™ Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.
Have Questions?
Catalog NumberType
A2-BPLarge, w/Buffer Exchange Ports
Catalog number A2-BP
Price (USD)
1,206.00
Online Exclusive
1,340.00
Save 134.00 (10%)
Each
Estimated availability date  Pending  
Add to cart
Type:
Large, w/Buffer Exchange Ports
Price (USD)
1,206.00
Online Exclusive
1,340.00
Save 134.00 (10%)
Each
Add to cart
Ask our AI about this Product

Handle high throughput with consistent, reliable results with Thermo Scientific™Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.

Features:

  • Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
  • Optional built-in buffer exchange ports allow for buffer recirculation on extended runs
  • UVT gel tray has gasketed end gates for leakproof seal without tape
  • Built-in leveling ensures flat, uniformly thick gels
  • Comb slots allow two, three or four sample sets of equal lengths to be run on the same gel
  • Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
  • Samples may be loaded directly from a 96-well plate, 8 or 12 at a time

Includes:

  • Buffer chamber with three-point leveling base
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • UVT gel tray
  • Gasketed end gates
  • Three combs (16-, 24- and 36-well), 1.5mm thick
Specifications
Certifications/ComplianceCE marked
DescriptionComplete system with buffer exchange ports
Dimensions (L x W x H)14.8 x 12.8 x 4.1 in. (37.5 x 32.5 x 10.5 cm)
Height (English)4.1 in.
Height (Metric)10.5 cm
Length (English)14.8 in.
Length (Metric)37.5 cm
Length (Metric) Gel25 cm
Sample Size8 to 432 (16-, 24- and 36-well combs)
Volume (Metric) Buffer2.3 L
Width (English)12.8 in.
Width (Metric)32.5 cm
Width (Metric) Gel20 cm
Product LineOwl
TypeLarge, w/Buffer Exchange Ports
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link