Gator A2 Large System 24-Slot Rapid Load UVT Gel Trays, with gasketed End Gates
Gator A2 Large System 24-Slot Rapid Load UVT Gel Trays, with gasketed End Gates
Actual product may vary
Thermo Scientific™
Gator A2 Large System 24-Slot Rapid Load UVT Gel Trays, with gasketed End Gates
Load multiple samples with Fisherbrand™ A2 and A5 Large Gel 24-Slot Rapid Load UVT Gel Trays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
A2-RL-2424 18-well Combs
A2-RL-1212 18-well Combs
Catalog number A2-RL-24
Price (USD)
-
Request A Quote
Includes:
24 18-well Combs
Ask our AI about this Product
Load multiple samples with Fisherbrand™ A2 and A5 Large Gel 24-Slot Rapid Load UVT Gel Trays.
Each side of UVT gel tray is fitted with a gasketed end gate which provides a leakproof seal without tape. Built-in leveling ensures the casting of flat, uniformly thick gels.
Specifications
Description24-slot rapid load UVT gel tray
For Use With (Equipment)Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems
TypeGel Tray
Includes24 18-well Combs
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection