GeneArt™ CRISPR Nuclease Vector with OFP Reporter Kit
GeneArt™ CRISPR Nuclease Vector with OFP Reporter Kit
Invitrogen™

GeneArt™ CRISPR Nuclease Vector with OFP Reporter Kit

The GeneArt™ CRISPR Nuclease Vector with OFP Reporter Kit is a vector system for expression of the functional components neededRead more
Have Questions?
Catalog NumberQuantity
A2117410 reactions
Catalog number A21174
Price (USD)
850.00
Each
-
Add to cart
Quantity:
10 reactions
Price (USD)
850.00
Each
Add to cart
The GeneArt™ CRISPR Nuclease Vector with OFP Reporter Kit is a vector system for expression of the functional components needed for CRISPR/Cas9 genome editing in mammalian cells with an orange fluorescent protein (OFP) reporter. The OFP reporter allows for fluorescence-based tracking of transfection efficiency, as well as FACS-based sorting/enrichment of Cas9 and CRISPR expressing cells. The linearized GeneArt™ CRISPR nuclease vectors provide a rapid and efficient way to clone double-stranded oligonucleotides encoding a crRNA representing a desired target into an expression cassette that allows sequence-specific targeting of the Cas9 nuclease. A version with competent cells is also available (Cat. No. A21178).

The GeneArt™ CRISPR Nuclease Vector system offers:
• Easy-to-design genome engineering system
• Affordable, ready-to-use cloning vectors
• Enrichment for hard to transfect or difficult to modify cell lines

All-in-one Vector System for CRISPR-based Genome Editing
The GeneArt™ CRISPR Nuclease Vector kit offers a simple, ready-to-use, all-in-one expression vector system consisting of both a Cas9 nuclease expression cassette and a guide RNA (gRNA) cloning cassette for rapid and efficient cloning of DNA that encodes target-specific CRISPR RNA (crRNA). This system allows you to edit and engineer a genomic locus of choice in a sequence-specific manner from a single plasmid. After relevant targets have been identified with fast and easy-to-use GeneArt™ CRISPR vectors, the biologically relevant mutations can be precisely created with GeneArt™ Precision TALs, with high specificity and low off-target effects.

Need assistance with CRISPR gRNA design?
Our new CRISPR Search & Design tool allows you to search our database of >600,000 predesigned CRISPR gRNAs in human and mouse genes or analyze your sequence of interest for de novo gRNA designs using our proprietary algorithms. CRISPR sequences are optimized for gene knockout and target the first three transcribed exons for each gene. Search results include the top 6 CRISPR sequences with PAM sites, exon maps with gRNA binding sites, and potential off-target binding sites for each CRISPR sequence. The tool will design the correct gRNA format for your preferred CRISPR-Cas9 product, including oligos for your GeneArt™ CRISPR Nuclease Vector. Start designing today >
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cloning MethodRestriction Enzyme/MCS
FormatKit
No. of Reactions10 Reactions
Overhang3' overhangs
Product LineGeneArt™
Product TypeCrispr Nuclease Vector With ofP Reporter Kit
PromoterU6, CMV
Protein TagOFP fusion
Quantity10 reactions
Reporter GeneOFP
Starting MaterialOligos (DNA)
Sufficient For10 Reactions
TechniqueCRISPR-Cas9
Detection MethodPrimer-probe
For Use With (Application)Gene Expression, Cloning
FormLiquid
Reaction SpeedFast
Unit SizeEach
Contents & Storage
Contains:
• CRISPR OFP Nuclease Vector, linearized
• 10X Oligonucleotide Annealing Buffer
• DNase/RNase-free Water
• 5X Ligation Buffer
• T4 DNA Ligase
• U6 Forward Sequencing Primer
• Control Double-stranded Oligonucleotide

Store in freezer (-5 to -30°C).

Frequently asked questions (FAQs)

What is CRISPR-STOP?

CRISPR-STOP is a method of inserting STOP codon sequences to generate knockouts.

Please refer to the following article: CRISPR-STOP: gene silencing through base-editing-induced nonsense mutations.

Find additional tips, troubleshooting help, and resources within our Genome Editing Support Center.

What transfection methods do you recommend when working with your CRISPR products?

For transfecting of the Cas9 protein, we would recommend using the Neon transfection system or Lipofectamine CRISPRMAX Cas9 Transfection Reagent. For transfection of mRNA, we would recommend using Lipofectamine MessengerMAX Transfection Reagent. For transfection of CRISPR vectors, we would recommend using Lipofectamine 3000 Transfection reagent.

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.

Do you have CRISPR products optimized for plants?

Our CRISPR products are optimized for mammalian systems, and have not been optimized for plant systems. However, we know researchers are doing that kind of work. While we have not tested our CRISPR products for plants, our new protein format would be ideal since it does not need to be translated and transcribed in the cell, so no plant-specific promoters required.

What are the benefits of using TAL/TALEN-based genome editing compared to your CRISPR system?

Invitrogen GeneArt Precision TALs, in addition to gene deletion, down-regulation and integration, can also be used for gene activation. Additionally, the system is based on a protein-DNA system, in contrast to CRISPR, which is based on a RNA-DNA system. TALs can be used to target any gene in any cell, including mammalian, bacterial, yeast, plants, insect, stem cells and zebrafish. Lastly, off-target effects are low when using the TAL system. Please refer to the following paper (http://www.sciencedirect.com/science/article/pii/S016816561500200X) where the authors compared TALs and CRISPR technology.

I am using the Invitrogen GeneArt CRISPR Nuclease Vector Kit and am getting very few ampicillin-resistant colonies on my selection plate. What could be the cause of this? Do you have any suggestions?

Here are possible reasons and solutions:

- Single-stranded (ss) oligonucleotides designed incorrectly; Make sure that each ss oligonucleotide contains the 5 nucleotides on the 3′ end required for cloning into the Invitrogen GeneArt CRISPR Nuclease Vector: ie., Top strand includes GTTTT on the 3′ end; Bottom strand includes CGGTG on the 3′ end.

- Double-stranded (ds) oligonucleotides were degraded; Store the 5 nM ds oligonucleotide stock in 1X Oligonucleotide Annealing Buffer. Avoid repeated freeze/thaw cycles; aliquot the 5 nM ds oligonucleotide stock and store at -20°C.

- Oligonucleotide annealing reaction inefficient; Ensure that the annealing reaction was performed as directed. If ambient temperature is >25°C, incubate the annealing reaction in a 25°C incubator.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link