Invitrogen™

Amplex™ Red Neuraminidase (Sialidase) Assay Kit

Catalog number: A22178
Have Questions?
Invitrogen™

Amplex™ Red Neuraminidase (Sialidase) Assay Kit

Catalog number: A22178
Have Questions?
Catalog Number
A22178
Unit Size
400 assays
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A22178400 assays
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Amplex™ Red Neuraminidase (Sialidase) Assay Kit provides a sensitive and simple method for detecting neuraminidase activity in serum and in purified enzyme systems using either a fluorescence microplate reader or fluorometer.

See our complete line of Fluorescence Microplate assays.

• Detect neuraminidase levels as low as 0.2 mU/mL
• Format allows for multiple time point measurements
• Designed for minimal autofluorescence interference
• Detect neuraminidase activity in biological samples such as serum

Neuraminidase (also known as sialidase) is a very common enzyme that hydrolyzes terminal sialic acid residues on polysaccharide chains, most often exposing a galactose residue. Neuraminidase plays a key role in the invasion of target cells and the subsequent replication of the influenza virus, making neuraminidase an important target for influenza drug development.

The Amplex™ Red Neuraminidase (Sialidase) Assay Kit provides an ultrasensitive method for detecting neuramimidase activity. This assay utilizes Amplex™ Red reagent to detect hydrogen peroxide generated by galactose oxidase oxidation of desialiated galactose, the end result of neuraminidase action. Then the hydrogen peroxide, in the presence of horseradish peroxidase (HRP), reacts in a 1:1 stoichiometric ratio with Amplex™ Red reagent to generate the red-fluorescent oxidation product resorufin.

Because resorufin has absorption and fluorescence emission maxima of approximately 571 nm and 585 nm, respectively, there is little interference from autofluorescence in most biological samples.

Use Amplex™ Red Assays for a Broad Range of Investigations
A wide variety of validated Amplex™ Red assays are available for studying cell signaling and lipids, neurobiology, inflammation and immune function, and metabolism. We also offer Amplex™ UltraRed Reagent (Cat. No. A36006), a second-generation reagent providing greater sensitivity and brighter fluorescence, and the Amplex™ Red/UltraRed Stop Reagent (Cat. No. A33855). The Amplex™ Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. After addition of the stop reagent, the fluorescence signal remains stable for at least three hours. Custom assay design and packaging are also available.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Fluorescent
For Use With (Application)
Neuraminidase (Sialidase) Assay
For Use With (Equipment)
Microplate Reader, Spectrophotometer, Fluorometer
Product Type
Red Neuraminidase (Sialidase) Assay Kit
Dye Type
Other Label(s) or Dye(s)
Format
Tube(s), 96-well plate
Product Line
Amplex™
Shipping Condition
Room Temperature

Contents & Storage

Store in freezer -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text