Invitrogen™

Amplex™ Red Galactose/Galactose Oxidase Assay Kit

Catalog number: A22179
Have Questions?
Invitrogen™

Amplex™ Red Galactose/Galactose Oxidase Assay Kit

Catalog number: A22179
Have Questions?
Catalog Number
A22179
Unit Size
400 assays
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A22179400 assays
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Amplex™ Red Galactose/Galactose Oxidase Assay Kit provides a sensitive and simple method for detecting galactose oxidase activity or galactose using a fluorescence microplate reader or fluorometer.

See our complete line of Fluorescence Microplate assays.

• Detect concentrations as low as 2 mU/mL of galactose oxidase activity or 4 μM galactose
• Format allows for multiple time point measurements
• Designed for minimal autofluorescence interference

Galactose oxidase is an important tool in the detection of galactose in biological fluids. The Amplex™ Red Galactose/Galactose Oxidase Assay Kit provides an ultrasensitive method for detecting galactose or for monitoring galactose oxidase activity. Galactose oxidase catalyzes the oxidation of galactose at the C6 position and generates hydrogen peroxide. In the presence of horseradish peroxidase (HRP), hydrogen peroxide reacts in a 1:1 stoichiometric ratio with the Amplex™ Red reagent to generate the red-fluorescent compound resorufin.

Because resorufin has absorption and fluorescence emission maxima of approximately 571 nm and 585 nm, respectively, there is little interference from autofluorescence in most biological samples.

Use Amplex™ Red Assays for a Broad Range of Investigations
A wide variety of validated Amplex™ Red assays are available for studying cell signaling and lipids, neurobiology, inflammation and immune function, and metabolism. We also offer Amplex™ UltraRed Reagent (Cat. No. A36006), a second-generation reagent providing greater sensitivity and brighter fluorescence, and the Amplex™ Red/UltraRed Stop Reagent (Cat. No. A33855). The Amplex™ Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. After addition of the stop reagent, the fluorescence signal remains stable for at least three hours. Custom assay design and packaging are also available.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Fluorescent
For Use With (Application)
Galactose/Galactose Oxidase Assay
For Use With (Equipment)
Microplate Reader, Spectrophotometer, Fluorometer
Product Type
Red Galactose/Galactose Oxidase Assay Kit
Dye Type
Other Label(s) or Dye(s)
Format
Tube(s), 96-well plate
Product Line
Amplex™
Shipping Condition
Room Temperature

Contents & Storage

Store in freezer -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text