Invitrogen™

Amplex™ Red Xanthine/Xanthine Oxidase Assay Kit

Catalog number: A22182
Have Questions?
Invitrogen™

Amplex™ Red Xanthine/Xanthine Oxidase Assay Kit

Catalog number: A22182
Have Questions?
Catalog Number
A22182
Unit Size
400 assays
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A22182400 assays
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Amplex Red Xanthine/Xanthine Oxidase Assay Kit provides a sensitive and simple fluorometric method for detecting as little as 200 nM xanthine or hypoxanthine or as little as 0.1 mU/mL of xanthine oxidase activity in a purified system with a 100 μL assay volume.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Fluorescent
For Use With (Application)
Xanthine/Xanthine Oxidase Assay
For Use With (Equipment)
Microplate Reader, Spectrophotometer, Fluorometer
Product Type
Red Xanthine/Xanthine Oxidase Assay Kit
Dye Type
Other Label(s) or Dye(s)
Format
Tube(s), 96-well plate
Product Line
Amplex™
Shipping Condition
Room Temperature

Contents & Storage

Store in freezer -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text