Invitrogen™

Alexa Fluor™ 350 Phalloidin

Catalog number: A22281
Have Questions?
Invitrogen™

Alexa Fluor™ 350 Phalloidin

Catalog number: A22281
Have Questions?
Catalog Number
A22281
Unit Size
300 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A22281300 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Alexa Fluor 350 phalloidin is a high-affinity F-actin probe conjugated to our bright, photostable, blue fluorescent Alexa Fluor 350 dye.

Selectively stains F-actin
Outstanding fluorescence performance
Excitation/Emission: 346/442 nm
Superior to antibody staining
Optimal for fixed and permeabilized samples

Get Superior Results in Actin Staining Studies
Phalloidin is a bicyclic peptide belonging to a family of toxins isolated from the deadly Amanita phalloides 'death cap' mushroom and is commonly used in imaging applications to selectively label F-actin. Fluorescently-labeled phalloidin has virtually identical binding properties with actin from different species including plants and animals. Phalloidin binds F-actin with high selectivity while Alexa Fluor 350 provides blue fluorescence of unparalleled brightness and photostability. Demonstrating very little nonspecific staining, Alexa Fluor 350 phalloidin allows high-contrast discrimination of actin staining.

Use in Multiple Applications
Alexa Fluor 350 phalloidin can be used to visualize and quantitate F-actin in tissue sections, cell cultures, or cell-free preparations. Alexa Fluor 350 phalloidin staining is fully compatible with other fluorescent stains used in cellular analyses including fluorescent proteins, Qdot nanocrystals, and other Alexa Fluor conjugates including secondary antibodies.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Links
Read about labeling F-Actin with phallotoxins.
See the full line of Alexa Fluor Dye products.
Create a Cellular Masterpiece with Alexa Fluor Phalloidin using the Cell Paint Tool.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Excitation Wavelength Range
346⁄442
Dye Type
Alexa Fluor™ 350
Product Line
Alexa Fluor™
Shipping Condition
Room Temperature
Label Type
Alexa Fluor Dyes
Product Type
Phalloidin
SubCellular Localization
Actin, Cytoskeleton

Contents & Storage

Store in freezer -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text