ViraPower™ BacMam Expression System
ViraPower™ BacMam Expression System
Invitrogen™

ViraPower™ BacMam Expression System

The ViraPower™ BacMam Expression System provides BacMam pCMV-Dest Vector, Cellfectin™ II Reagent, and MAX Efficiency™ DH10Bac™ Competent Cells for easyRead more
Have Questions?
Catalog NumberQuantity
A242271 Kit
Catalog number A24227
Price (USD)
1,888.00
Each
Add to cart
Quantity:
1 Kit
Price (USD)
1,888.00
Each
Add to cart
Ask our AI about this Product

The ViraPower™ BacMam Expression System provides BacMam pCMV-Dest Vector, Cellfectin™ II Reagent, and MAX Efficiency™ DH10Bac™ Competent Cells for easy recombination-based cloning and baculovirus-based expression of a target gene. With BacMam technology, a modified insect cell virus (baculovirus) is used as a vehicle to efficiently deliver and transiently express genes in mammalian cells with minimum effort and toxicity. This technology allows for control over the level of expression through the increase or decrease of viral particle concentration.When combined with Gateway™ cloning technology, a variety of sizes of target genes can be cloned and expressed. Sizes ranging from an insert for RNAi to a large gene with introns (38 kb) have been cloned and expressed successfully using the BacMam pCMV-Dest Vector. This plasmid accommodates various cloning schemes, including Gateway™, Seamless Assembly, and traditional cloning using restriction enzymes.

The BacMam pCMV-Dest Vector contains a CMV promoter to drive constitutive expression of the target gene. It also contains VSV-G elements, which enable baculovirus to have high transduction efficiency, and a WPRE element, which elongates transient expression in mammalian systems. To facilitate successful cloning, AmpR and gentamicin have been incorporated for positive selection, as well as Cm(R) (chloramphenicol resistance gene) for negative selection. Upon successful cloning, the region containing the CmR is replaced by the insert, selecting against any bacteria that still have a CmR-containing region.

Cellfectin™ II Reagent (included) is a cationic-lipid formulation designed for optimal transfection of insect cells. The system also includes MAX Efficiency™ DH10Bac™ Competent Cells, which are optimized for the production of recombinant baculovirus molecules for the expression of eukaryotic proteins.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Delivery TypeBaculovirus (BacMam System)
For Use With (Application)Viral Expression, Constitutive Expression, Multi-Gene Expression
Product TypeInsect Cell Expression System
Quantity1 Kit
Reporter GeneNone
Selection Agent (Eukaryotic)None
VectorpDEST
Cloning MethodRestriction Enzyme/MCS, Seamless Cloning, Gateway
Concentration500 ng/μL
Product LineViraPower™
PromoterCMV
Protein TagUntagged
Unit SizeEach
Contents & Storage

• BacMam pCMV-Dest Vector, 10 μg, store at 2°C to 8°C
• MAX Efficiency™ DH10Bac™ Chemically Competent Cells, 0.5 mL, store at -68°C to -85°C
• Cellfectin™ Reagent, 1 mL, store at 2°C to 8°C

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection