Lysis Buffer 1 + RNase for Food ID
Lysis Buffer 1 + RNase for Food ID
Thermo Scientific™
Lysis Buffer 1 + RNase for Food ID
This convenient packaging of Lysis Buffer 1 + RNase for Food ID is recommended for use with the PrepSEQ NucleicWeitere Informationen
Have Questions?
KatalognummerMenge
A244014 x 0,5 l
Katalognummer A24401
Preis (EUR)
293,00
Each
Geschätzte Verfügbarkeit 21-Jul-2025
Zum Warenkorb hinzufügen
Menge:
4 x 0,5 l
Preis (EUR)
293,00
Each
Zum Warenkorb hinzufügen
Ask our AI about this Product
This convenient packaging of Lysis Buffer 1 + RNase for Food ID is recommended for use with the PrepSEQ Nucleic Acid Extraction Kit (Cat. No. 4480466) to prepare DNA for meat ID and GMO testing from food and feed samples. The protocol using Lysis Buffer 1 + RNase in conjunction with the PrepSEQ Nucleic Acid Extraction Kit can be readily automated on the Thermo Scientific KingFisher mL Purification System platform to reduce hands-on time and increase reproducibility of the DNA extraction procedure. The purified DNA can then be used in PCR or other molecular biology techniques, and is well-suited for use with the entire line of RapidFinder™ Species ID Kits and TaqMan™ GMO Kits.
Specifications
Hochdurchsatz-KompatibilitätGeeignet für hohe Durchsätze
Menge4 x 0,5 l
ProbentypLebensmittel- und Umweltproben
Unit SizeEach
Inhalt und Lagerung
Enthält:
• 4 Flaschen Lysepuffer 1
• Je 2 Röhrchen RNase, getrocknet
Bei Raumtemperatur lagern.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Dokumente und Downloads
Zertifikate
Sicherheitsdatenblätter
Geben Sie die Katalognummer, den Namen oder Link frei.
1x1 image pixel for data collection