Neurobasal™-A Medium, no D-glucose, no sodium pyruvate
Neurobasal-A medium without D-glucose and sodium pyruvate is used to support the growth of post-natal CNS neurons in applications whereRead more
Have Questions?
Catalog number A2477501
Price (USD)/ 500 mL
111.65
Online exclusive
119.00
Save 7.35 (6%)
-
Add to cart
Price (USD)/ 500 mL
111.65
Online exclusive
119.00
Save 7.35 (6%)
Add to cart
Neurobasal™-A Medium, no D-glucose, no sodium pyruvate
Catalog numberA2477501
Price (USD)/ 500 mL
111.65
Online exclusive
119.00
Save 7.35 (6%)
-
Add to cart
Neurobasal-A medium without D-glucose and sodium pyruvate is used to support the growth of post-natal CNS neurons in applications where the presence of D-glucose and sodium pyruvate can cause interference.
For neural studies that require the absence of insulin, glucose, and sodium pyruvate, such as glucose response or insulin response studies, we recommend Gibco B-27 Supplement Minus Insulin as a supplement to this medium.
Neurobasal Medium is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations of Neurobasal media and B-27 supplements.