Applied Biosystems™
Precision Plate Holder for 7500 Real-Time PCR Systems
Catalog number: A24820
Have Questions?
Applied Biosystems™
Precision Plate Holder for 7500 Real-Time PCR Systems
Catalog number: A24820
Have Questions?
Catalog Number
A24820
Unit Size
1 each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A248201 each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This precision plate holder is required for use of MicroAmp® EnduraPlate™ optical reaction plates on Applied Biosystems® 7500 real-time PCR systems. It also acommodates MicroAmp® optical reaction plates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)
7500 System
Product Type
Precision Plate Holder
For Use With (Application)
Real Time PCR (qPCR)
Quantity
1 Ea.
PCR Method
qPCR
Contents & Storage
7500 Reactions