SILAC RPMI 1640 Flex Media, no glucose, no phenol red
SILAC RPMI 1640 Flex Media, no glucose, no phenol red
Gibco™

SILAC RPMI 1640 Flex Media, no glucose, no phenol red

SILAC RPMI 1640 Medium is RPMI 1640 basal cell culture medium without L-arginine, L-glutamine, and L-lysine for use during SILACRead more
Have Questions?
Catalog number A2494201
Price (USD)/ 500 mL
88.65
Online exclusive
90.00 
Save 1.35 (2%)
-
Add to cart
Request bulk or custom format
Price (USD)/ 500 mL
88.65
Online exclusive
90.00 
Save 1.35 (2%)
Add to cart
SILAC RPMI 1640 Flex Media, no glucose, no phenol red
Catalog numberA2494201
Price (USD)/ 500 mL
88.65
Online exclusive
90.00 
Save 1.35 (2%)
-
Add to cart
SILAC RPMI 1640 Medium is RPMI 1640 basal cell culture medium without L-arginine, L-glutamine, and L-lysine for use during SILAC protein labeling with stable isotope-labeled lysine and/or arginine. RPMI 1640 Medium is widely used for supporting the growth of many different mammalian cells. Cells successfully cultured in RPMI-1640 include HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of Gibco™ RPMI 1640 modifications for a range of applications. Find the right modification using the media selector tool.
This RPMI 1640 Medium is modified as follows:
WithWithout
 • Glucose
 • Phenol red
 • HEPES
 • L-arginine
 • L-glutamine
 • L-lysine

RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 Medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 Medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). SILAC protein labeling experiments should be conducted using Gibco™ Dialyzed FBS. RPMI 1640 Medium uses a sodium bicarbonate buffer system (2.0 g/L), and therefore requires a 5-10% CO2 environment to maintain physiological pH.

cGMP Manufacturing and Quality System
SILAC RPMI 1640 Medium is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Culture TypeMammalian Cell Culture
FormLiquid
Product TypeRPMI 1640
Serum LevelStandard Serum Supplementation
Shelf Life12 Months
Without AdditivesNo Glucose, No Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate, No Arginine, No Lysine
Green FeaturesSustainable packaging
Product LineGibco™
Quantity500 mL
Unit Size500 mL
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.
Shelf life: 12 months from date of manufacture.