Precision ID Identity Panel
Precision ID Identity Panel
Applied Biosystems™

Precision ID Identity Panel

The Applied Biosystems Precision ID Identity Panel is one of the first next-generation sequencing (NGS) solutions for human identification thatRead more
Have Questions?
Catalog NumberQuantity
A2564396 reactions
Catalog number A25643
Price (EUR)
1.294,00
Each
-
Add to cart
Quantity:
96 reactions
Recurring order eligible. Learn more »
Price (EUR)
1.294,00
Each
Add to cart
Ask our AI about this Product
The Applied Biosystems Precision ID Identity Panel is one of the first next-generation sequencing (NGS) solutions for human identification that uniquely identifies pristine or degraded samples. The multiplexed panel of SNPs provides discrimination of individuals similar to STR genotype match probabilities used by forensic analysts (between 1x10-31 and 6x10-35).

High discrimination power is achieved by using 34 upper Y-Clade SNPs and 90 autosomal SNPs that have high heterozygosity and low fixation index (Fst) as described in publications by Dr. Kenneth Kidd from Yale University and the SNPForID Consortium (1-3). The panel design has been optimized for use with degraded or challenging forensic samples using only 1 ng of starting DNA input, although acceptable performance has been observed with sub-nanogram input quantities as low as 100 pg. The autosomal markers have an average amplicon length of 132 nucleotides and the Y-markers are 141 nucleotides in length on average. Using a small sample input of as little as 1 ng of DNA, you can go from sample extraction to genotypes in less than a two days using the Precision ID NGS System and Converge Software for analysis.

Simplicity
• Small amplicons enable better recovery with degraded samples
• As little as 100 pg of input
• Automated library and template preparation using the Ion Chef System

Scalability
• Multiple chip formats to meet a range of sample throughput requirements
• Ability to multiplex up to 384 samples*

Speed
• Less than 45 minutes of hands-on time for a DNA-to-data targeted sequencing workflow*
• Total sample-to-data time of less than 2 days*

NOTE: The Precision ID Identity Panel provides sufficient material to perform 96 reactions using the manual Precision ID Library kit (Cat. No. A26435) or 32 reactions using the automated Precision ID DL8 Library Kit (Cat. No. A33212).

* Using the Precision ID NGS System for Human Identification

References:
1. Pakstis, AJ, Speed, WC, Fang, R, Hyland, FC, Furtado, MR, Kidd, JR, and Kidd, KK. (2010) SNPs for a universal individual identification panel. Human Genetics 127(3), 315-324.
2. Phillips, C, Fang, R, Ballard, D, Fondevila, M, Harrison, C, Hyland, F, et al. (2007) Evaluation of the Genplex SNP typing system and a 49plex forensic marker panel. Forensic Science International: Genetics 1(2), 180-185.
3. Karafet, TM, Mendez, FL, Meilerman, MB, Underhill, PA, Zegura, SL, and Hammer, MF. (2008) New binary polymorphisms reshape and increase resolution of the human Y chromosomal haplogroup tree. Genome Research 18(5), 830-838.

Specifications
Final Product TypePCR Amplicons
For Use With (Application)Sequencing
For Use With (Equipment)Ion GeneStudio S5 System, Ion PGM™ System, Ion S5™ XL System, Ion OneTouch™ 2 System, Ion Chef™ System
LibrariesTargeted Sequencing Library
No. of Reactions96 reactions
Product LineIon AmpliSeq™, Ion Torrent™
Product TypePrecision ID Identity Panel
Quantity96 reactions
Sample TypeDNA
SpeciesHuman
Workflow StepLibrary Generation
Sequencing TypeGenome and DNA Sequencing
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2407017Certificate of AnalysisSep 19, 2024A25643
2112016Certificate of AnalysisMay 30, 2022A25643
2103015Certificate of AnalysisSep 06, 2021A25643
1810014Certificate of AnalysisFeb 19, 2020A25643
4 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection