Informational:Informational:Experience nuclei counting with fluorescent-mode counting algorithms, simplified workflows, built-in dilution calculators and reports using the latest version of Countess software with optional 21 CFR Part 11 compliance. Download now!
Invitrogen™

Countess™ Reusable Slide

Catalog number: A25750
Have Questions?
Invitrogen™

Countess™ Reusable Slide

Catalog number: A25750
Have Questions?
Catalog Number
A25750
Unit Size
1 unit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A257501 unit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
For use with the Countess II FL, Countess 3, and Countess 3 FL automated cell counters. The Countess Reusable Slide is a greener alternative to disposable slides.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Reusable Slide
Product Line
Countess™
For Use With (Application)
Cell Counting
For Use With (Equipment)
Countess II FL, Countess 3, and 3 FL Automated Cell Counters
Shipping Condition
Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text