Applied Biosystems™

GeneScan™ 600 LIZ™ Size Standard v2.0 Dx

Catalog number: A25794
Have Questions?
Applied Biosystems™

GeneScan™ 600 LIZ™ Size Standard v2.0 Dx

Catalog number: A25794
Have Questions?
Catalog Number
A25794
Unit Size
800 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A25794800 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The GeneScan™ 600 LIZ™ Size Standard v2.0 Dx is a five dye–labeled, high-density size standard for the reproducible sizing of fragment analysis data. This product can be used as a traditional size standard on all of the Applied Biosystems™ capillary electrophoresis systems. Use this size standard for fragments between 20 and 600 bp. The standard contains 36 LIZ dye–labeled, single-stranded DNA fragments. Since the standard is labeled with the fifth dye, users can genotype a greater number of markers in a given lane, compared to the four-dye system.

Size fragments in the 20–600 nucleotides range
GeneScan 600 LIZ Size Standard v2.0 Dx is designed for sizing DNA fragments in the 20–600 nucleotides range and provides 36 single-stranded, labeled fragments of: 20, 40, 60, 80, 100, 114, 120, 140, 160, 180, 200, 214, 220, 240, 250, 260, 280, 300, 314, 320, 340, 360, 380, 400, 414, 420, 440, 460, 480, 500, 514, 520, 540, 560, 580, and 600. The sizing curve generated from fragments make the GeneScan 600 LIZ Size Standard v2.0 Dx ideal for use in a variety of fragment analysis applications such as analysis of microsatellites and fragment length polymorphisms as well as in relative fluorescent quantitation. Each of the DNA fragments is labeled with the LIZ fluorophore, which results in a single peak when run under denaturing conditions. With the fifth dye, LIZ, your marker fragments can be labeled with FAM™, VIC™, NED™, or PET™ dye.

Specifications

Format
Standard
For Use With (Equipment)
3500 Dx Genetic Analyzer
GC-Rich PCR Performance
High
For Use With (Application)
Sequencing
Label or Dye
LIZ
No. of Reactions
800 Reactions
Product Line
GeneScan™, LIZ™
Product Type
DNA Sequencing System
Shipping Condition
Wet Ice
Size
20 bp, 600 bp
Test Time
Fast

Contents & Storage

Store at 2°C to 8°C. Protect from light. Do not freeze.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text