Discover our new Invitrogen E-Gel Agarose Gels with SYBR Safe DNA Stain—a safe alternative to ethidium bromide. Learn more »
This product is experiencing limited availability.
Invitrogen™
E-Gel™ NGS™ 0.8% Agarose Gels
E-Gel NGS 0.8% Agarose Gels are self-contained, pre-cast agarose gels designed to provide fast, convenient, and easy electrophoresis. Each E-GelRead more
Have Questions?
Catalog number A25798
Price (USD)/ 10 gels
211.00
Add to cart
Price (USD)/ 10 gels
211.00
Add to cart
E-Gel™ NGS™ 0.8% Agarose Gels
Catalog numberA25798
Price (USD)/ 10 gels
211.00
Add to cart
E-Gel NGS 0.8% Agarose Gels are self-contained, pre-cast agarose gels designed to provide fast, convenient, and easy electrophoresis. Each E-Gel agarose gel is ready for use with agarose, electrodes, and SYBR Safe DNA gel stain packaged inside an 11-well, easy-to-open, UV- and blue-light transparent cassette. SYBR Safe stain can be viewed with blue light, which minimizes DNA damage and allows safe, real-time monitoring of DNA migration using the E-Gel Power Snap Electrophoresis System.
Easy access to the gel The gel cassette is designed to open with a gel knife, so that the gel inside can be accessed readily for excision of specific bands or transferred to a membrane for Southern blot analysis.