Applied Biosystems™

resDNASEQ™ Human Residual DNA Quantitation Kits

Catalog number: A27335
Have Questions?
Applied Biosystems™

resDNASEQ™ Human Residual DNA Quantitation Kits

Catalog number: A27335
Have Questions?
The resDNASEQ Human Residual DNA Quantitation Kit is a quantitative PCR (qPCR)-based system for the detection of host cell DNA from human cell lines used as expression systems for recombinant proteins. Reliable and rapid, the resDNASEQ system enables sensitive (as low as 15 pg DNA/mL test sample) and specific quantitation of human cell DNA. This performance helps ensure a high degree of confidence in quantitation data obtained from a broad range of sample types, from in-process samples to final product, whether the sample contains high molecular weight or sheared DNA.
 
Catalog Number
A27335
Unit Size
100 reactions
Includes
Assay Kit and Sample Preparation
Price (USD)
Availability
-
Full specifications
Quantity
Detection MethodPrimer-probe
FormatKit
PCR MethodqPCR
Host CellHuman DNA
Label or DyeFAM
Product LineresDNASEQ™
No. of Reactions100 Reactions
IncludesAssay Kit and Sample Preparation
Unit Size100 reactions
Catalog Number
Unit Size
100 reactions
Includes
Assay Kit only
Price (USD)
Availability
-
Full specifications
Quantity
Detection MethodPrimer-probe
FormatKit
PCR MethodqPCR
Host CellHuman DNA
Label or DyeFAM
Product LineresDNASEQ™
Shipping ConditionDry Ice
No. of Reactions100 Reactions
IncludesAssay Kit only
Unit Size100 reactions
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeIncludesPrice (USD)AvailabilityQuantity
A27335Full specifications
100 reactionsAssay Kit and Sample Preparation
-
Detection MethodPrimer-probe
FormatKit
PCR MethodqPCR
Host CellHuman DNA
Label or DyeFAM
Product LineresDNASEQ™
No. of Reactions100 Reactions
IncludesAssay Kit and Sample Preparation
Unit Size100 reactions
A26366Full specifications
100 reactionsAssay Kit only
-
Detection MethodPrimer-probe
FormatKit
PCR MethodqPCR
Host CellHuman DNA
Label or DyeFAM
Product LineresDNASEQ™
Shipping ConditionDry Ice
No. of Reactions100 Reactions
IncludesAssay Kit only
Unit Size100 reactions
Showing 2 of 2
Features of the resDNASEQ Human Residual DNA Quantitation Kit include:
• Highly sensitive quantitation using proven TaqMan real-time qPCR technology
• Consistent performance across the expected range of DNA fragment sizes
• Includes master mix, TaqMan primer/probe mix, and human genomic DNA standard

This kit is available with or without sample preparation reagents in the form of a PrepSEQ Residual DNA Sample Preparation Kit, which is optimized for highly efficient residual DNA recovery from complex mixtures of proteins, buffers, and salts.

The resDNASEQ Human Residual DNA Quantitation Kit provides highly sensitive detection of human DNA, allowing you to use small sample volumes to generate accurate results. The broad linear range of TaqMan real-time qPCR technology allows you to test samples with variable levels of human DNA in the same assay, such as in-process samples with higher amounts of DNA or bulk drug substance with very low amounts.

Easy to use
With the resDNASEQ Human Residual DNA Quantitation Kit you perform qPCR analysis to compare the amount of human DNA in your test samples to a standard curve generated with known amounts of purified human genomic DNA standard. The resDNASEQ kit includes almost everything you need to carry out these steps, from master mix to TaqMan primer/probe mix to human genomic DNA standard, as well as step-by-step instructions.

Consistent performance
For accurate quantitation of residual human DNA, assay results must be unaffected by the size of the DNA molecules present in the test sample. With the resDNASEQ system you get consistent assay performance across a range of DNA fragment sizes.

*Due to the high degree of similarity between genomes, DNA from closely-related primate species such as monkey (Vero), chimpanzee, and orangutan are expected be detected by the resDNASEQ Human Quantitation Kit.

Contents & Storage

resDNASEQ Human DNA Control
• Human DNA Control, 30 ng/μL, 40 μL, store at –15 to –25°C
• DNA Dilution Buffer (DDB), 7 mL, store at –15 to –25°C before first use and 2−8°C after first use

resDNASEQ Human DNA Real-Time PCR Reagents
• Environmental Master Mix (2X), 2 x 0.75 mL, store at –15 to –25°C before first use and 2−8°C after first use, protect from light
• Human DNA Assay Mix (10X), 300 μL, store at –15 to –25°C, protect from light
• Negative Control (water), 1.0 mL, store at –15 to –25°C before first use and 2−8°C after first use

PrepSEQ Nucleic Acid Extraction Kit
• Lysis Buffer (2 bottles, 50 mL/bottle)
• Binding Solution/Isopropanol (1 empty bottle)
• Wash Buffer Concentrate (2 bottles, 26 mL/bottle)
• Elution Buffer (1 bottle, 25 mL)
• Proteinase K Buffer (1 bottle, 50 mL)
• Magnetic Particles (2 tubes, 1.5 mL/tube)
• Proteinase K (20 mg/mL) (1 tube, 1.25 mL), store at or below –20°C
• PK Buffer II (1 bottle, 11 mL)
Store all at room temperature unless otherwise indicated.

PrepSEQ Residual DNA Sample Preparation Kit
• Proteinase K (20 mg/mL), 1 tube, 1.25 mL
• Yeast tRNA (10 mg/mL), 1 tube, 0.5 mL
• Glycogen (5 mg/mL), 2 tubes, 1.0 mL/tube
Store all at or below –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text